MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Separation of amino acids by capillary electrophoresis with light-emitting diode-induced fluorescence in the presence of electroosmotic flow.
Separation of amino acids by capillary electrophoresis with light-emitting diode-induced fluorescence in the presence of electroosmotic flow.
Methods in molecular biology (Clifton, N.J.) (2013-02-07)
Ming-Mu Hsieh, Po-Ling Chang
ABSTRACT
In this chapter, we describe a method to identify amino acids (AA) by capillary electrophoresis in conjunction with light-emitting diode-induced fluorescence (LEDIF). First, amino acids labeled with naphthalene-2,3-dicarboxaldehyde (NDA) in the presence of cyanide are converted to highly fluorescent cyanobenz[f]isoindole (CBI) derivatives. Next, they are separated by gel electrophoresis in the presence of EOF. In the process, the CBI products were excited by a violet LED to produce green fluorescence. In addition to the optical setup of light-emitting diode-induced fluorescence, the preparation of poly(ethylene) oxide for amino acid separation is also described in this chapter.
Sigma-Aldrich
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence