MilliporeSigma
Aqueous extraction of pectin from sisal waste.
Carbohydrate polymers (2013-02-13)
Jener David G Santos, Alexandre F Espeleta, Alexsandro Branco, Sandra A de Assis
ABSTRACT
In this work, sisal waste was used as a source of pectin. Sisal is known worldwide as a source of hard fibres, and Brazil is the largest producer of sisal, producing more than 246,000 tonnes. However, the process of removing the fibres of the sisal leaf generates 95% waste. This study investigated the effect of the liquid/solid ratio (%), time (min), and temperature (°C) on the yield of the pectin obtained from sisal waste by attractive environmentally friendly process. A statistical Box-Behnken design was applied to determine the important effects and interactions of these independent variables on the yield of pectin, the dependent variable. Significant models were obtained. The yield of the extracted pectin ranged from 4.61 to 19.2%. The conditions that produced the highest yield (19.2%) were a temperature of 85 °C, extraction time of 60 min and a liquid/solid ratio of 2%.
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Supelco
Pectin, for use with Total Dietary Fiber Control Kit, TDF-C10