MilliporeSigma
Blood media for the cultivation of Mycobacterium tuberculosis.
American journal of clinical pathology (1951-02-01)
M S TARSHIS, A W FRISCH
Product Description
Millipore
Blood Agar (Base), NutriSelect® Plus, powder, for bacteria, pack of 100 g and 500 g, suitable for microbiology