MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Silver-zwitterion organic-inorganic nanocomposite with antimicrobial and antiadhesive capabilities.
Silver-zwitterion organic-inorganic nanocomposite with antimicrobial and antiadhesive capabilities.
Langmuir : the ACS journal of surfaces and colloids (2013-02-22)
Rong Hu, Guozhu Li, Yujiao Jiang, Yi Zhang, Ji-Jun Zou, Li Wang, Xiangwen Zhang
ABSTRACT
In this work, we demonstrate a convenient, efficient, and environmentally benign strategy to achieving antimicrobial and antiadhesive purposes using a silver-zwitterion nanocomposite. The synthesis of the nanocomposite relies on loading zwitterionic polymer brushes with Ag(+) precursor ions, followed by their in situ reduction to Ag nanoparticle by ultraviolet (UV) irradiation. Both poly(sulfobetaine methacrylate) (pSBMA) and poly(carboxybetaine methacrylate) (pCBMA) have been studied as matrices for the embedding of silver. Well-dispersed silver nanoparticles are embedded into pCBMA matrices. The obtained pCBMA-silver hybrid (CB-Ag) is capable of killing bacteria upon contact and releasing dead bacteria under wet conditions. Results suggest the feasibility of using this nanocomposite system as a robust and reliable antimicrobial and antiadhesive platform for the prevention of microbial colonization.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent