MilliporeSigma
ECG of the month. Multiple episodes of collapse in a cat.
Journal of the American Veterinary Medical Association (2013-03-23)
Yamir Reina Doreste, Joshua A Stern, Sunshine M Lahmers
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Supelco
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Diltiazem hydrochloride solution, 1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Diltiazem hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Diltiazem for system suitability, European Pharmacopoeia (EP) Reference Standard