MilliporeSigma
  • Efficacy of triclosan-based toothpastes in the prevention and treatment of plaque-induced periodontal and peri-implant diseases.
Efficacy of triclosan-based toothpastes in the prevention and treatment of plaque-induced periodontal and peri-implant diseases.
Minerva stomatologica (2013-03-23)
L Trombelli, R Farina
ABSTRACT
To evaluate the efficacy of triclosan (T)-based toothpaste formulations in the prevention and treatment of plaque-induced periodontal and peri-implant diseases. A review of the existing literature was conducted with a systematic approach in order to retrieve pertinent articles. i) Compared with a control fluoride dentifrice, a fluoride dentifrice containing T formulations provides a more effective level of plaque control and gingival health in patients affected by gingivitis; ii) 0.3% T/2% copolymer/0.243% NaF formulation and 0.3% T/0.13% Ca glicerophosphate/1000 ppm F toothpaste in a natural Ca carbonate base seem the most effective T-based toothpaste formulations in controlling plaque and gingival inflammation in patients with gingivitis or mild/moderate periodontitis over a 6-month period; iii) 0.3% T/2% copolymer/0.243% NaF toothpaste formulation can reduce clinical attachment loss in young adolescents when compared with a 0.243% NaF toothpaste formulation, the magnitude of the difference being greater for patients with deep periodontal pockets at baseline; iv) 0.3% T/2% copolymer/0.243% NaF toothpaste formulation is either similarly or more efficacious in preventing the progression/recurrence of periodontal destruction when compared to a conventional fluoride toothpaste; v) 0.3% T/2% copolymer/0.243% NaF toothpaste formulation seems to be more effective than a fluoride toothpaste formulation in controlling the severity of mucosal inflammation, the incidence of mucosal bleeding as well as reducing probing pocket depth around dental implants.
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material