MilliporeSigma
  • Effect of buffer systems and disaccharides concentration on Podoviridae coliphage stability during freeze drying and storage.
Effect of buffer systems and disaccharides concentration on Podoviridae coliphage stability during freeze drying and storage.
Cryobiology (2013-03-30)
C Dini, P J de Urraza
ABSTRACT
The aims of this study were to determine the stability of Podoviridae coliphage CA933P during lyophilization and storage in different media, and to establish similarities between the results obtained and those expected through mechanisms described for proteins stabilization during freeze-drying. PBS and SM buffer were assayed as lyophilization media. The effect of inorganic salts concentration as well as the addition of disaccharides on phage stability during freeze-drying and storage was also studied. The addition of low sucrose concentration (0.1 mol l⁻¹) to SM buffer stabilized phage during freezing and drying steps of the lyophilization process, but higher sugar concentrations were detrimental to phage stability during freeze-drying. Sucrose stabilized phage during storage for at least 120 days. The lyoprotective effect of low concentrations of disaccharides during the drying step of the lyophilization of proteins as well as the stabilization of the freeze-dried product in time correlated with the results obtained for phage CA933P.
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sucrose, European Pharmacopoeia (EP) Reference Standard