MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Estimation of usual intake and food sources of choline and betaine in New Zealand reproductive age women.
Estimation of usual intake and food sources of choline and betaine in New Zealand reproductive age women.
Asia Pacific journal of clinical nutrition (2013-05-03)
Vanessa L Mygind, Sophie E Evans, Meredith C Peddie, Jody C Miller, Lisa A Houghton
ABSTRACT
Recently, choline has been associated with neurodevelopment, cognitive function and neural tube defect incidence. However, data on usual intakes are limited, and estimates of dietary intakes of choline and its metabolite betaine, are not available for New Zealanders. The objective of the present study was to determine usual intake and food sources of choline and betaine in a group of New Zealand reproductive age women. Dietary intake data were collected from a sample of 125 women, aged 18-40 years, by means of a 3-day weighed food record, and usual choline and betaine intake distributions were determined. The mean (SD) daily intakes of choline and betaine were 316 (66) mg and 178 (66) mg, respectively. The total choline intake relative to energy intake and body weight was 0.18 mg/kcal and 5.1 mg/kg, respectively. Only 16% of participants met or exceeded the Adequate Intake (AI) for adult women of 425 mg of choline. The top five major food contributors of choline were eggs, red meat, milk, bread and chicken; and of betaine were bread, breakfast cereal, pasta, grains and root vegetables (carrots, parsnips, beetroot, swedes). Our findings contribute towards the recent emergence of published reports on the range of dietary choline and betaine intakes consumed by free-living populations. In our sample of New Zealand women, few participants were meeting or exceeding the AI level. Given recent epidemiological evidence suggesting health benefits of increased choline and betaine intakes, recommendations should be made to encourage the consumption of choline and betaine-rich foods.
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent