MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Adenosine production by human B cells and B cell-mediated suppression of activated T cells.
Adenosine production by human B cells and B cell-mediated suppression of activated T cells.
Blood (2013-05-17)
Zenichiro Saze, Patrick J Schuler, Chang-Sook Hong, Dongmei Cheng, Edwin K Jackson, Theresa L Whiteside
ABSTRACT
Antibody-independent role of B cells in modulating T-cell responses is incompletely understood. Freshly isolated or cultured B cells isolated from the peripheral blood of 30 normal donors were evaluated for CD39 and CD73 coexpression, the ability to produce adenosine 5'-monophosphate (AMP) and adenosine (ADO) in the presence of exogenous adenosine triphosphate (ATP) as well as A₁, A2A, A2B, and A₃ adenosine receptor (ADOR) expression. Human circulating B cells coexpress ectonucleotidases CD39 and CD73, hydrolyze exogenous ATP to 5'-AMP and ADO, and express messenger RNA for A₁R, A2AR, and A₃R. 2-chloroadenosine inhibited B-cell proliferation and cytokine expression, and only A₃R selective antagonist restored B-cell functions. This suggested that B cells use the A₃R for autocrine signaling and self-regulation. Mediated effects on B-cell growth ± ADOR antagonists or agonists were tested in carboxyfluorescein diacetate succinimidyl ester assays. In cocultures, resting B cells upregulated functions of CD4⁺ and CD8⁺ T cells. However, in vitro-activated B cells downregulated CD73 expression, mainly produced 5'-AMP, and inhibited T-cell proliferation and cytokine production. These B cells acquire the ability to restrict potentially harmful effects of activated T cells. Thus, B cells emerge as a key regulatory component of T cell-B cell interactions, and their dual regulatory activity is mediated by the products of ATP hydrolysis, 5'-AMP, and ADO.
Product Description
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder