MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • [Study on in situ intestinal absorption of baicalin contained in Tiangou Jiangya capsules].
[Study on in situ intestinal absorption of baicalin contained in Tiangou Jiangya capsules].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-31)
Ling-Jun Li, Ji Li, Hua-Wei Lou
ABSTRACT
To study in situ intestinal absorption kinetics of baicalin contained in Tiangou Jiangya capsules, and the effect of different intestinal segments, pH value, drug concentration and P-gp inhibitor on the absorption. The in situ intestinal perfusion test was adopted, and HPLC method was used to determine the content of baicalin in samples at different time points. Ultra-violet (UV) spectrophotometry was used to determine the content of phenol red in samples at different time points. When pH value was at 5. 0, 6. 5, 7. 4, the absorption of baicalin was not impacted. P-gp inhibitor verapamil could enhance the absorption of baicalin. When the quality concentration of the test solution ranged between 5-20 g L -1 , the linearity of the absorption amount of baicalin increased. The absorption kinetic equation of baicalin was Y = -0. 073 7X +0. 118 7 (r = 0. 994 8) , K. 0. 073 7 h -1 , t1/2 9. 40 h. Baicalin is mainly absorbed in colon. The absorption of baicalin shows the first-order kinetics process, with the absorption mechanism of passive diffusion. Baicalin is a substrate for P-gp.
Product Description
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Baicalin, 95%
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard