MilliporeSigma
  • [Regulation of osteogenic differentiation of mesenchimal stem sells of bone marrow].
[Regulation of osteogenic differentiation of mesenchimal stem sells of bone marrow].
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova (2013-07-19)
Iu M Zakharov, E B Makarova
ABSTRACT
The review describes the mechanisms of hormonal, autocrine, paracrine and intracellular regulation in osteogenic differentiation of mesenchymal stem cells, the role of negative feedbacks in this process.
Sigma-Aldrich
Fibroblast Growth Factor from bovine pituitary, suitable for cell culture