MilliporeSigma
  • Specificity of endoproteinase Asp-N (Pseudomonas fragi): cleavage at glutamyl residues in two proteins.
Specificity of endoproteinase Asp-N (Pseudomonas fragi): cleavage at glutamyl residues in two proteins.
Biochemical and biophysical research communications (1989-08-15)
D Ingrosso, A V Fowler, J Bleibaum, S Clarke
ABSTRACT
Endoproteinase Asp-N, a metalloprotease from a mutant strain of Pseudomonas fragi, has been reported to specifically cleave on the N-terminal side of aspartyl and cysteic acid residues. We utilized this enzyme to generate fragments for determining the amino acid sequence of the D-aspartyl/L-isoaspartyl methyltransferase isozyme I from human erythrocytes. Surprisingly, we identified cleavage sites for this enzyme at the N-terminal side of several glutamyl residues in addition to the expected cleavage sites at aspartyl residues. The ability of this enzyme to cleave polypeptides at both glutamyl and aspartyl residues was confirmed by mapping additional sites on erythrocyte carbonic anhydrase I. These results indicate that a more appropriate name for this enzyme may be Endoproteinase Asp/Glu-N.
Sigma-Aldrich
Endoproteinase Asp-N from Pseudomonas fragi mutant strain, suitable for protein sequencing, lyophilized powder