MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of the methyl esters of pyruvate, succinate and glutamate on the secretory response to meglitinide analogues in rat pancreatic islets.
Effects of the methyl esters of pyruvate, succinate and glutamate on the secretory response to meglitinide analogues in rat pancreatic islets.
Pharmacological research (1996-03-01)
A Bakkali Nadi, T M Zhang, W J Malaisse
ABSTRACT
The insulinotropic action of the meglitinide analogues KAD-1229, A-4166 and repaglinide was examined in rat pancreatic islets deprived of exogenous nutrient or incubated in the presence of nutrient secretagogues such as D-glucose and the methyl esters of pyruvic acid, succinic acid and glutamic acid. The meglitinide analogues exerted little effect on insulin release in the absence of exogenous nutrient or in the presence of methyl pyruvate. They caused obvious stimulation of insulin output in the presence of D-glucose, dimethyl succinate or dimethyl glutamate. It is proposed, therefore, that suitable esters of dicarboxylic nutrients could be used to potentiate the secretory response to meglitinide analogues in non-insulin-dependent diabetes mellitus.
Sigma-Aldrich
L-Glutamic acid dimethyl ester hydrochloride, ≥99.0% (anhydrous basis material, AT)
Sigma-Aldrich
Dimethyl succinate, 98%, FG
Sigma-Aldrich
DBE-4 dibasic ester, 98%