MilliporeSigma
  • Synthesis of cyclic and acyclic beta-amino acids via chelation-controlled 1,3-dipolar cycloaddition.
Synthesis of cyclic and acyclic beta-amino acids via chelation-controlled 1,3-dipolar cycloaddition.
The Journal of organic chemistry (2003-10-25)
Roger Hanselmann, Jiacheng Zhou, Philip Ma, Pat N Confalone
ABSTRACT
Isoxazolidines have been synthesized in diastereomeric excess up to 94% via a MgBr2-induced chelation-controlled 1,3-dipolar cycloaddition reaction with N-hydroxyphenylglycinol as a chiral auxiliary. The diastereomerically pure isoxazolidines were further transformed into cyclic and acyclic beta-amino acid derivatives.
Sigma-Aldrich
N-Phenylglycine, 97%