MilliporeSigma
  • Contribution of neural and endothelial isoforms of the nitric oxide synthase to the inhibitory effects of NO on the cat carotid body.
Contribution of neural and endothelial isoforms of the nitric oxide synthase to the inhibitory effects of NO on the cat carotid body.
Advances in experimental medicine and biology (2003-11-26)
Viviana Valdes, Matias Mosqueira, Rodrigo Iturriaga
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole