MilliporeSigma
Food-drug interactions: grapefruit juice.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (2011-06-21)
Camelia Harapu Diaconu, Magdalena Cuciureanu, L Vlase, Rodica Cuciureanu
ABSTRACT
Food-drug interactions are increasingly recognized as important clinical events which may change significantly the bioavailability of oral administrated drugs. Grapefruit juice (GFJ) demonstrated multiple interactions with drugs leading to loss of the therapeutic effects or increased side-effects. GFJ decreases pre-systemic metabolism through a) competitive or mechanism-based inhibition of gut wall CYP3A4 isoenzymes and b) P-glycoprotein (P-gp), c) multidrug resistance protein-2 (MRP2) or d) organic anion-transporting polypeptide (OATP) inhibition. Although, GFJ presents high amounts of flavonoids (e.g. naringin, naringenin), furanocoumarins (e.g. 6',7'-dihydroxybergamottin, bergamottin) are the main chemicals involved in the pharmacokinetic interactions. As compounds of GFJ show additive or synergistic effects, all the major furanocoumarins are necessary for the maximal inhibitory effect. Also, related citrus fruits (sweeties, pummelo and sour orange) or various plants containing furanocoumarins may present pharmacological interactions, yet to be discovered.
Product Description
Supelco
Bergamottin, analytical standard