MilliporeSigma
  • Mechanisms of oral carcinogenesis induced by dibenzo[a,l]pyrene: an environmental pollutant and a tobacco smoke constituent.
Mechanisms of oral carcinogenesis induced by dibenzo[a,l]pyrene: an environmental pollutant and a tobacco smoke constituent.
International journal of cancer (2013-03-14)
Kun-Ming Chen, Joseph B Guttenplan, Shang-Min Zhang, Cesar Aliaga, Timothy K Cooper, Yuan-Wan Sun, Joseph DelTondo, Wieslawa Kosinska, Arun K Sharma, Kun Jiang, Richard Bruggeman, Kwangmi Ahn, Shantu Amin, Karam El-Bayoumy
ABSTRACT
We previously reported that dibenzo[a,l]pyrene (DB[a,l]P), the most potent known environmental carcinogen among polycyclic aromatic hydrocarbons (PAH) congeners, is carcinogenic in the oral tissues of mice. We have now developed a new mouse model which employs the oral application of the fjord region diol epoxide, (±)-anti-11,12-dihydroxy-13,14-epoxy-11,12,13,14-tetrahydrodibenzo[a,l]pyrene (DB[a,l]PDE), a metabolite of the tobacco smoke constituent DB[a,l]P, and we show its specific induction of oral squamous cell carcinoma (OSCC) in both tongue and other oral tissues. Groups of B6C3F1 mice (20/group) received 6 or 3 nmol of (±)-anti-DB[a,l]PDE administered into the oral cavity; 3 times per week for 38 weeks. Additional groups received the vehicle alone or were left untreated. Mice were sacrificed 42 weeks after the first carcinogen administration. The high dose induced 74 and 100% OSCC in the tongue and other oral tissues, respectively; the corresponding values at the lower dose were 45 and 89%. Using immunohistochemistry, we showed that DB[a,l]PDE resulted in overexpression of p53 and COX-2 proteins in malignant tissues when compared to normal oral tissues and tongues. Consistent with the carcinogenicity, we demonstrated powerful mutagenicity in cII gene in B6C3F1 (Big Blue) mouse tongue. The mutational profile in lacI reporter gene is similar to those detected in human head and neck cancer, and p53 mutations were observed in mouse oral tumor tissues. Taken together, we conclude that the formation of diol epoxides plays a major role among the mechanisms by which DB[a,l]P exerts its oral mutagenicity and tumorigenicity.
Product Description
Dibenzo[a,l]pyrene, BCR®, certified reference material
Supelco
Dibenzo[a,l]pyrene, vial of 25 mg, Cerilliant®