MilliporeSigma
  • Functional impact of ABCB1 variants on interactions between P-glycoprotein and methadone.
Functional impact of ABCB1 variants on interactions between P-glycoprotein and methadone.
PloS one (2013-03-26)
Chin-Chuan Hung, Mu-Han Chiou, Yu-Ning Teng, Yow-Wen Hsieh, Chieh-Liang Huang, Hsien-Yuan Lane
ABSTRACT
Methadone is a widely used substitution therapy for opioid addiction. Large inter-individual variability has been observed in methadone maintenance dosages and P-glycoprotein (P-gp) was considered to be one of the major contributors. To investigate the mechanism of P-gp's interaction with methadone, as well as the effect of genetic variants on the interaction, Flp-In™-293 cells stably transfected with various genotypes of human P-gp were established in the present study. The RNA and protein expression levels of human P-gp were confirmed by real-time quantitative RT-PCR and western blot, respectively. Utilizing rhodamine 123 efflux assay and calcein-AM uptake study, methadone was demonstrated to be an inhibitor of wild-type human P-gp via non-competitive kinetic (IC50 = 2.17±0.10 µM), while the variant-type human P-gp, P-gp with 1236T-2677T-3435T genotype and P-gp with 1236T-2677A-3435T genotype, showed less inhibition potency (IC50 = 2.97±0.09 µM and 4.43±1.10 µM, respectively) via uncompetitive kinetics. Methadone also stimulated P-gp ATPase and inhibited verapamil-stimulated P-gp ATPase activity under therapeutic concentrations. These results may provide a possible explanation for higher methadone dosage requirements in patients carrying variant-type of P-gp and revealed the possible drug-drug interactions in patients who receive concomitant drugs which are also P-gp substrates.
Product Description
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder