MilliporeSigma
  • Amino acids form strongly bound anions when substituted with superhalogen ligands.
Amino acids form strongly bound anions when substituted with superhalogen ligands.
The Journal of chemical physics (2013-04-12)
Iwona Sieradzan, Iwona Anusiewicz
ABSTRACT
The properties of AA-Y(-) anions (where AA = cysteine, aspartic acid, lysine; Y = BF3, PF5) were investigated at the ab initio Outer Valence Green's Function (OVGF)∕6-311++G(3df,3pd)∕∕MP2∕6-311++G(d,p) level of theory. It is shown that introducing a superhalogen-like substituent to an amino acid (i.e., Cys, Asp, and Lys) results in obtaining molecules that bind an excess electron relatively strongly. The electronic stabilities of such resulting daughter anions are predicted to be substantial (5.3-6.9 eV).
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)