MilliporeSigma
  • Effect of blood product medium supplements on the activity of cefotaxime and other cephalosporins against Enterococcus faecalis.
Effect of blood product medium supplements on the activity of cefotaxime and other cephalosporins against Enterococcus faecalis.
Diagnostic microbiology and infectious disease (1989-03-01)
G M Eliopoulos, E Reiszner, S Willey, W J Novick, R C Moellering
ABSTRACT
The activities of cefotaxime and other aminothiazoyl oxime cephalosporins against Enterococcus faecalis were enhanced by addition of 5% sheep blood to Mueller-Hinton agar. This effect was not seen with aztreonam (aminothiazoyl oxime monobactam), cefotiam (aminothiazoyl, nonoxime), or other cephalosporins, and it was specific to the syn-configuration of the oxime moiety. Enhancement of cefotaxime activity was demonstrable against approximately 50% of 86 clinical isolates and could only be shown at low bacterial inocula. Human serum, serum alpha 1-, beta- and gamma-globulin fractions and albumin often antagonized or did not affect significantly the antimicrobial activity of cefotaxime, while the alpha 2-globulin fraction usually enhanced drug activity. The in vivo activity of cefotaxime against E. faecalis was examined in a rat peritoneal abscess model. The test organism was resistant to cefotaxime by standard methods (MIC greater than 128 micrograms/ml) but was inhibited by 1.0 microgram/ml when rat serum was presented in the medium. Cefotaxime reduced titers of bacteria within abscesses after 5 days of therapy (5.77 +/- 0.68 log10 CFU/g) in comparison with those in control animals (7.38 +/- 0.28 log10 CFU/g, p less than 0.05). Moxalactam, the in vitro activity of which was not augmented by serum, proved ineffective in the animal model. While these observations do not have direct therapeutic relevance, they offer a possible explanation for the relatively infrequent occurrence of enterococcal superinfection in patients treated with cefotaxime.
Product Description
Millipore
Mueller Hinton Agar, NutriSelect® Plus, according to FDA, NCCLS, powder, for bacteria, pack of 500 g, pack of 100 g, pack of 2.5 kg