MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Benzoate treatment and the glycine index in nonketotic hyperglycinaemia.
Benzoate treatment and the glycine index in nonketotic hyperglycinaemia.
Journal of inherited metabolic disease (2005-09-10)
J L K Van Hove, K Vande Kerckhove, J B Hennermann, V Mahieu, P Declercq, S Mertens, M De Becker, P S Kishnani, J Jaeken
ABSTRACT
High-dose benzoate treatment aimed at reducing plasma glycine levels to normal reduces seizures and increases wakefulness in patients with nonketotic hyperglycinaemia (NKH). Since benzoate metabolism is dependent on the available glycine pool, and since the glycine pool is variably affected by the deficiency in the glycine cleavage enzyme system, we examined the importance of interpatient variability in benzoate requirement. To correct for the dietary glycine contribution, the glycine index was introduced as the molar requirement of benzoate dose necessary to normalize plasma glycine levels and subtracting from that the dietary glycine intake, both corrected for weight. The glycine index varied between 3.62 and 4.87 mmol/kg per day in five patients with a poor neurodevelopmental outcome and between 0.92 and 1.90 mmol/kg per day in four patients with a better neurodevelopmental outcome, and was 2.54 mmol/kg per day in a single patient with an intermediate outcome. The glycine index was stable over time within each patient. Exceeding the balance by either increasing food glycine intake or decreasing the benzoate dose resulted in increased glycine levels. Exceeding the glycine tolerance by increasing benzoate resulted in elevated and toxic levels of benzoate. The glycine index is a stable, individually specific parameter in patients with NKH. It has clinical consequences for the dose of benzoate required and the role of dietary management. Through its correlation with neurodevelopmental outcome, the glycine index points to potential genetic factors that could contribute to the psychomotor retardation in NKH.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%