MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Ion-pairing reversed-phase liquid chromatography fractionation in combination with isotope labeling reversed-phase liquid chromatography-mass spectrometry for comprehensive metabolome profiling.
Ion-pairing reversed-phase liquid chromatography fractionation in combination with isotope labeling reversed-phase liquid chromatography-mass spectrometry for comprehensive metabolome profiling.
Journal of chromatography. A (2011-05-06)
Kevin Guo, Jun Peng, Ruokun Zhou, Liang Li
ABSTRACT
We report a novel two-dimensional (2D) separation strategy aimed at improving the detectability of liquid chromatography mass spectrometry (LC-MS) for metabolome analysis. It is based on the use of ion-pairing (IP) reversed-phase (RP) LC as the first dimension separation to fractionate the metabolites, followed by isotope labeling of individual fractions using dansylation chemistry to alter the physiochemical properties of the metabolites. The labeled metabolites having different hydrophobicity from their unlabeled counterparts are then separated and analyzed by on-line RPLC Fourier-transform ion-cyclotron resonance mass spectrometry (FTICR-MS). This off-line 2D-LC-MS strategy offers significant improvement over the one-dimensional (1D) RPLC MS technique in terms of the number of detectable metabolites. As an example, in the analysis of a human urine sample, 3564 ¹³C-/¹²C-dansylated ion pairs or metabolites were detected from seven IP RPLC fractions, compared to 1218 metabolites found in 1D-RPLC-MS. Using a library of 220 amine- and phenol-containing metabolite standards, 167 metabolites were positively identified based on retention time and accurate mass matches, which was about 2.5 times the number metabolites identified by 1D-RPLC-MS analysis of the same urine sample.
Product Description
Supelco
Hydrochlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrochlorothiazide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Sigma-Aldrich
2-Phenylglycine, 95%
Sigma-Aldrich
Hydrochlorothiazide, crystalline
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%
Protocatechuic acid, primary reference standard
Sigma-Aldrich
DL-2-Aminocaprylic acid, 99%
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Hydrochlorothiazide, meets USP testing specifications
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
DL-o-Tyrosine, ≥96.0% (NT)
Sigma-Aldrich
o-Cresol, ≥99%
Supelco
Diethanolamine, analytical standard
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
2-Aminoisobutyric acid, 98%
Supelco
o-Cresol, analytical standard
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethanolamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Diethanolamine, JIS special grade, ≥99.0%