MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • High fetal estrogen concentrations: correlation with increased adult sexual activity and decreased aggression in male mice.
High fetal estrogen concentrations: correlation with increased adult sexual activity and decreased aggression in male mice.
Science (New York, N.Y.) (1983-06-17)
F S vom Saal, W M Grant, C W McMullen, K S Laves
ABSTRACT
In the house mouse (Mus musculus), fetuses may develop in utero next to siblings of the same or opposite sex. The amniotic fluid of the female fetuses contains higher concentrations of estradiol than that of male fetuses. Male fetuses that developed in utero between female fetuses had higher concentrations of estradiol in their amniotic fluid than males that were located between other male fetuses during intrauterine development. They were also more sexually active as adults, less aggressive, and had smaller seminal vesicles than males that had developed between other male fetuses in utero. These findings raise the possibility that during fetal life circulating estrogens may interact with circulating androgens both in regulating the development of sex differences between males and females and in producing variation in phenotype among males and among females.
Product Description
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®