MilliporeSigma
  • Identification of electrophysiologically-active compounds for the malaria mosquito, Anopheles gambiae, in human sweat extracts.
Identification of electrophysiologically-active compounds for the malaria mosquito, Anopheles gambiae, in human sweat extracts.
Medical and veterinary entomology (1996-07-01)
A Cork, K C Park
ABSTRACT
Human sweat samples were chemically fractionated into acid and non-acid components. The most abundant volatile compounds present in the fractions were identified by linked gas chromatography mass spectrometry. The acid fractions were found to be composed of a range of twenty aliphatic and three aromatic carboxylic acids ranging, on average, from 0.02 to 20 micrograms per ml of sweat sampled. Non-acid fractions were found to contain: 6-methyl-5-hepten-2-one, 1-octen-3-ol, decanal, benzyl alcohol, dimethylsulphone, phenylethanol, phenol and 4-methylphenol, collectively amounting to 0.1 and 3 micrograms per ml of sweat. The major component of sweat was found to be L-lactic acid which constituted from 1 to 5 mg/ml. Using the intact antennae of the anthropophilic malaria vector mosquito Anopheles gambiae Giles, the peripheral olfactory activities of compounds identified in the sweat fractions were investigated by electroantennography (EAG). Short-chain saturated carboxylic acids, methanoic, ethanoic, propanoic, butanoic, pentanoic and hexanoic acids were found to elicit significantly larger EAG responses than longer chain saturated carboxylic acids from female An.gambiae. For a given dose the largest amplitude EAG response was elicited by methanoic acid. Pentanoic acid elicited larger EAG responses than either butanoic or hexanoic acids. Two non-acidic compounds, 1-octen-3-ol and 4-methylphenol, were found to elicit significant dose-dependent EAG responses from female An.gambiae. 1-Octen-3-ol elicited larger EAG responses than 4-methylphenol for a given dose, but both compounds elicited smaller EAG responses than the same dose of C1-C6 straight-chain aliphatic carboxylic acids. The possible behavioural significance of the EAG-active compounds identified in human sweat samples is discussed.
Sigma-Aldrich
p-Cresol, JIS special grade, ≥99.0%
Supelco
4-Methylphenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Ethyl heptanoate, ReagentPlus®, 99%
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
p-Cresol, 99%
Sigma-Aldrich
Ethyl heptanoate, ≥98%, FG
Sigma-Aldrich
Ethyl heptanoate, natural, ≥98%, FG
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%