MilliporeSigma
  • Hyphenation of ultra performance liquid chromatography (UPLC) with inductively coupled plasma mass spectrometry (ICP-MS) for fast analysis of bromine containing preservatives.
Hyphenation of ultra performance liquid chromatography (UPLC) with inductively coupled plasma mass spectrometry (ICP-MS) for fast analysis of bromine containing preservatives.
Journal of pharmaceutical and biomedical analysis (2006-01-19)
Lars Bendahl, Steen Honoré Hansen, Bente Gammelgaard, Stefan Sturup, Camilla Nielsen
ABSTRACT
Ultra performance liquid chromatography (UPLC) was coupled to inductively coupled plasma mass spectrometry (ICP-MS) for fast analysis of three bromine-containing preservatives, monitoring the 79Br and 81Br isotopes simultaneously. Due to the efficiency of the 1.7 microm column packing material, the resolution of the test substances was only slightly affected when the linear flow velocity was increased from 0.5 to 1.9 mm s(-1). However, the sensitivity of ICP-MS detection decreased when the linear flow velocity was increased from 0.5 to 1.9 mm s(-1). Analytical figures of merit were determined at an intermediate and at a high linear velocity. The precision was better than 2.2% R.S.D. and regression analysis showed that a linear response was achieved at both flow rates (R2 > 0.9993, n = 36). The analysis time was less than 4.5 min at a flow rate of 50 microL min(-1) and limits of detection and quantification were better than 3.3 and 11 microg BrL(-1), respectively. The analysis time was reduced to 2.7 min when the flow rate was increased to 90 microL min(-1) and limits of detection and quantification were better than 20 and 65 microg BrL(-1), respectively. The method was applied for quantitative analysis of bromine-containing preservatives in commercially available cosmetic products.
Product Description
Supelco
Bronopol, PESTANAL®, analytical standard
Supelco
Methyldibromoglutaronitrile, analytical standard