MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A daptomycin-xylitol-loaded polymethylmethacrylate bone cement: how much xylitol should be used?
A daptomycin-xylitol-loaded polymethylmethacrylate bone cement: how much xylitol should be used?
Clinical orthopaedics and related research (2013-03-27)
Ali Salehi, Ashley Cox Parker, Gladius Lewis, Harry S Courtney, Warren O Haggard
ABSTRACT
The rate of release of an antibiotic from an antibiotic-loaded polymethylmethacrylate (PMMA) bone cement is low. This may be increased by adding a particulate poragen (eg, xylitol) to the cement powder. However, the appropriate poragen amount is unclear. We explored the appropriate amount of xylitol to use in a PMMA bone cement loaded with daptomycin and xylitol. We prepared four groups of cement, each comprising the same amount of daptomycin in the powder (1.36 g/40 g dry powder) but different amounts of xylitol (0, 0.7, 1.4, and 2.7 g); the xylitol mass ratio (X) (mass divided by mass of the final dry cement-daptomycin-xylitol mixture) ranged from 0 to 6.13 wt/wt%. Eight mechanical, antibiotic release, and bacterial inhibitory properties were determined using three to 22 specimens or replicates per test. We then used an optimization method to determine an appropriate value of X by (1) identifying the best-fit relationship between the value of each property and X, (2) defining a master objective function incorporating all of the best fits; and (3) determining the value of X at the maximum master objective function. We found an appropriate xylitol amount to be 4.46 wt/wt% (equivalent to 1.93 g xylitol mixed with 1.36 g daptomycin and 40 g dry cement powder). We demonstrated a method that may be used to determine an appropriate xylitol amount for a daptomycin-xylitol-loaded PMMA bone cement. These findings will require in vivo confirmation. While we identified an appropriate amount of xylitol in a daptomycin-xylitol-loaded PMMA bone cement as a prophylactic agent in total joint arthroplasties, clinical evaluations are needed to confirm the effectiveness of this cement.
Sigma-Aldrich
Daptomycin, cyclic lipopeptide antibiotic
Supelco
Xylitol, analytical standard
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Sigma-Aldrich
Xylitol, ≥99% (GC)