MilliporeSigma
  • Chronic social stress during adolescence: interplay of paroxetine treatment and ageing.
Chronic social stress during adolescence: interplay of paroxetine treatment and ageing.
Neuropharmacology (2013-04-23)
Sebastian H Scharf, Vera Sterlemann, Claudia Liebl, Marianne B Müller, Mathias V Schmidt
ABSTRACT
Exposure to chronic stress during developmental periods is a risk factor for a number of psychiatric disorders. While the direct effects of stress exposure have been studied extensively, little is known about the long-lasting effects and the interaction with ageing. The same holds true for the treatment with selective serotonin reuptake inhibitors (SSRIs), which have been shown to prevent or reverse some stress-induced effects. Here, we studied the direct and long-lasting impact of chronic social stress during adolescence and the impact of chronic treatment with the SSRI paroxetine in adulthood and aged animals. Therefore, male CD1 mice at the age of 28 days were subjected to 7 weeks of chronic social stress. Treatment with paroxetine was performed per os with a dosage of 20 mg/g BW. We were able to reverse most of the effects of chronic social stress in adult mice (4 months old) and to some extend in aged animals (15 months old) with the SSRI treatment. Especially the regulation of the HPA axis seems to be affected in aged mice with a shift to the use of vasopressin. Our results demonstrate that chronic stress exposure and antidepressant treatment at the end of the developmental period can have a significant and long-lasting impact, highly relevant for healthy ageing.
Sigma-Aldrich
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 IU/mL in 0.9% NaCl
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Corticotropin Releasing Factor sheep, ≥95% (HPLC)
Sigma-Aldrich
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid