MilliporeSigma
  • Reduction in lipid oxidation by incorporation of encapsulated sodium tripolyphosphate in ground turkey.
Reduction in lipid oxidation by incorporation of encapsulated sodium tripolyphosphate in ground turkey.
Meat science (2013-06-12)
Marsha L Sickler, James R Claus, Norman G Marriott, William N Eigel, Hengjian Wang
ABSTRACT
Ground turkey, with 1% NaCl, was incorporated with no sodium tripolyphosphate (control, nSTP), unencapsulated STP (uSTP; 0.3% or 0.5%), encapsulated STP (eSTP; 0.3% or 0.5% active, phosphate basis), or a blend (0.3% uSTP plus 0.2% eSTP). Encapsulate (hydrogenated vegetable oil) was designed to melt at 74°C. Treatments were stored (4, 24h at 3°C) before being cooked to two different endpoints (EPT; 74, 79°C) followed by post-cooked storage (0, 5, 10days). An improvement of 77% (0.3% eSTP) and 80% (0.5% eSTP) in the reduction of TBARS was found in comparison to corresponding uSTP. The blend produced a 62% improvement compared to uSTP (0.5%) while maintaining cook yield. CIE a* values were highest at both EPT and post-cooked storage times beyond 0 day for eSTP. Meat manufacturing procedures that entail a delayed thermal processing step will benefit by an improvement in lipid oxidation control through the use of encapsulated phosphates.
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%