MilliporeSigma
  • Changes of placental syndecan-1 expression in preeclampsia and HELLP syndrome.
Changes of placental syndecan-1 expression in preeclampsia and HELLP syndrome.
Virchows Archiv : an international journal of pathology (2013-06-29)
Szilvia Szabo, Yi Xu, Roberto Romero, Tibor Fule, Katalin Karaszi, Gaurav Bhatti, Tibor Varkonyi, Ildiko Varkonyi, Tibor Krenacs, Zhong Dong, Adi L Tarca, Tinnakorn Chaiworapongsa, Sonia S Hassan, Zoltan Papp, Ilona Kovalszky, Nandor Gabor Than
ABSTRACT
Preeclampsia is characterized by maternal systemic anti-angiogenic and pro-inflammatory states. Syndecan-1 is a cell surface proteoglycan expressed by the syncytiotrophoblast, which plays an important role in angiogenesis and resolution of inflammation. Our aim was to examine placental syndecan-1 expression in preeclampsia with or without hemolysis, elevated liver enzymes, and low platelet count (HELLP) syndrome. Placentas were obtained from women in the following groups: (1) late-onset preeclampsia (n = 8); (2) early-onset preeclampsia without (n = 7) and (3) with HELLP syndrome (n = 8); (4) preterm controls (n = 5); and (5) term controls (n = 9). Tissue microarrays (TMAs) were constructed from paraffin-embedded placentas. TMA slides were immunostained for syndecan-1 and evaluated using microscopy, virtual microscopy, and semi-automated image analysis. Maternal sera from patients with preeclampsia (n = 49) and controls (n = 32) were immunoassayed for syndecan-1. BeWo cells were treated with Forskolin or Latrunculin B or kept in ischemic conditions. SDC1 expression and syndecan-1 production were investigated with qRT-PCR, confocal microscopy, and immunoassays. Syndecan-1 was localized to the syncytiotrophoblast apical membrane in normal placentas. Syndecan-1 immunoscores were higher in late-onset preeclampsia (p = 0.0001) and early-onset preeclampsia with or without HELLP syndrome (p = 0.02 for both) than in controls. Maternal serum syndecan-1 concentration was lower in preeclampsia (median, 673 ng/ml; interquartile range, 459-1,161 ng/ml) than in controls (1,158 ng/ml; 622-1,480 ng/ml). SDC1 expression and syndecan-1 immunostainings in BeWo cells and syndecan-1 concentrations in supernatants increased during cell differentiation. Disruption of the actin cytoskeleton with Latrunculin B decreased syndecan-1 release, while ischemic conditions increased it. Syncytiotrophoblastic syndecan-1 expression depends on the differentiation of villous trophoblasts, and trophoblastic syndecan-1 release is decreased in preeclampsia and HELLP syndrome. This phenomenon may be related to the disturbed syncytiotrophoblastic cortical actin cytoskeleton and associated with maternal anti-angiogenic and pro-inflammatory states in these syndromes.
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder