MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Trichloroethylene adsorption by pine needle biochars produced at various pyrolysis temperatures.
Trichloroethylene adsorption by pine needle biochars produced at various pyrolysis temperatures.
Bioresource technology (2013-07-11)
Mahtab Ahmad, Sang Soo Lee, Anushka Upamali Rajapaksha, Meththika Vithanage, Ming Zhang, Ju Sik Cho, Sung-Eun Lee, Yong Sik Ok
ABSTRACT
In this study, pine needles were converted to biochar (BC) at different pyrolysis temperatures of 300, 500, and 700 °C to sorb trichloroethylene (TCE), and the changes in BC properties with each temperature were evaluated. Pyrolysis temperature showed a pronounced effect on BC properties. Decreases in molar H/C and O/C ratios resulted from removing O- and H-containing functional groups with increasing temperature, and produced high aromaticity and low polarity BCs. BCs produced at higher temperature showed greater TCE removal efficiency from water due to their high surface area, micro-porosity, and carbonized extent. The performance of various BCs for TCE removal was assessed by the Freundlich, Langmuir, Temkin, and Dubinin-Radushkevich adsorption models, among which the Temkin and Dubinin-Radushkevich models best described TCE adsorption onto various BCs, indicating prevailing sorption mechanism as pore-filling.
Product Description
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichloroethylene, JIS special grade, ≥99.5%