MilliporeSigma
  • Strongyloides cebus (Nematoda: Strongyloididae) in Lagothrix cana (Primates: Atelidae) from the Brazilian Amazon: aspects of clinical presentation, anatomopathology, treatment, and parasitic biology.
Strongyloides cebus (Nematoda: Strongyloididae) in Lagothrix cana (Primates: Atelidae) from the Brazilian Amazon: aspects of clinical presentation, anatomopathology, treatment, and parasitic biology.
The Journal of parasitology (2013-08-06)
Vitor Luís Tenório Mati, Francisco Carlos Ferreira Junior, Hudson Alves Pinto, Alan Lane de Melo
ABSTRACT
Abstract :  Seven cases of parasitism by Strongyloides cebus were identified in Lagothrix cana from Brazil. Aspects of the clinical presentation, treatment, pathology, and parasitic biology of these infections are described. Moderate to severe disease was observed, requiring hospitalization of 3 primates, and diarrhea was the most common clinical sign described. One L. cana individual died, for which ulcerative enteritis was the major finding upon histopathological analysis. The use of ivermectin in these atelids was safe and effective against the parasite. Parallel attempts to experimentally infect gerbils with the parasite failed. Lagothrix cana is presented as a new host for S. cebus. The evidence that Strongyloides infections are common in nonhuman primates under free-living conditions, and even more prevalent in captive animals, likely represents a neglected problem.
Product Description
Ivermectin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ivermectin