MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A novel indole compound, AWT-489, inhibits prostaglandin D2-induced CD55 expression by acting on DP prostanoid receptors as an antagonist in LS174T human colon cancer cells.
A novel indole compound, AWT-489, inhibits prostaglandin D2-induced CD55 expression by acting on DP prostanoid receptors as an antagonist in LS174T human colon cancer cells.
Archives of biochemistry and biophysics (2013-11-19)
Satomi Oyama, Hiromichi Fujino, Risa Yamazaki, Iori Okura, John W Regan, Atsuko Awata, Takayoshi Arai, Toshihiko Murayama
ABSTRACT
Indoles are composed of a common core structure, the indole ring, and are widely used as pharmaceuticals and their precursors. In this study, a newly composed relatively small indole compound, AWT-489 was examined to find a novel specific antagonist for DP receptors; the cognate receptors for prostaglandin D2 (PGD2), to prevent colon cancer malignancy. Here we showed that AWT-489 antagonized DP receptor-mediated cyclic AMP formation, and expression of CD55, an inhibitor of the complement system that correlates with poor survival in patients with colorectal cancer, in LS174T human colon cancer cells. Interestingly, unlike a popular indole compound, indomethacin, AWT-489 did not act on the cyclooxygenases as a non-steroidal anti-inflammatory drug. Moreover, AWT-489 exhibited a better inhibitory effect than that of the well-used DP receptor antagonist, BWA868C when a dose close to the physiological concentration of PGD2 was used. These results suggest that AWT-489 can act as a novel human DP receptor antagonist to reduce the expression of CD55 in LS174T human colon cancer cells. We believe that AWT-489 has potential as a lead compound for designing a new DP receptor antagonist that may help improve PGD2-related diseases, especially colon cancer in the near future.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin D2, ≥95%, synthetic
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Dinoprostone, European Pharmacopoeia (EP) Reference Standard