MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • 8-hydroxydeoxyguanosine as a useful marker for determining the severity of trichloroethylene exposure.
8-hydroxydeoxyguanosine as a useful marker for determining the severity of trichloroethylene exposure.
Archives of environmental & occupational health (2013-12-12)
Sedat Abusoglu, Huseyin Tugrul Celik, Engin Tutkun, Hinç Yilmaz, Muhittin A Serdar, Ceylan Demir Bal, M Yildirimkaya, M Avcikucuk
ABSTRACT
The aim of this study was to determine 8-hydroxydeoxyguanosine (8-OH-dG) levels in trichloroethylene (TCE)-exposed workers. Oxidative stress biomarkers and biochemical parameters were monitored among 26 TCE-exposed workers and 78 age-matched control subjects. Levels of urinary 8-OH-dG were analyzed by liquid chromatography tandem mass spectrometry. Urinary 8-OH-dG levels were significantly higher for TCE-exposed group (p < .001). Spearman's correlation test revealed positive correlations between urinary trichloroacetic acid levels and age, urinary 8-OH-dG, urinary total oxidant status, and urinary total antioxidant status (p = .042, p < .001, p < .001, and p < .001, respectively). 8-OH-dG may be a useful marker to determine the extent of TCE exposure.
Product Description
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Trichloroethylene, JIS special grade, ≥99.5%
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis