MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • [Protective effect of isoorientin on alcohol-induced hepatic fibrosis in rats].
[Protective effect of isoorientin on alcohol-induced hepatic fibrosis in rats].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2014-02-06)
Yong-Xin Chen, Quan-Fang Huang, Xing Lin, Jin-Bin Wei
ABSTRACT
To observe the effect and mechanism of isoorientin from Gypsophila elegans on alcohol-induced hepatic fibrosis in rats. ninety healthy male Wistar rats were randomly divided into six groups: the normal control group, the model control group, the colchicines group (positive control, 1.0 mg x kg(-1) x d(-1)), the high, middle and low-dose isoorientin groups (20, 50, 100 mg x kg(-1) x d(-1)). The normal control group received normal saline, while other groups received alcohol to cause hepatic fibrosis. After 24-weeks treatment, the alanine aminotransferase (ALT), aspartate aminotransferase (AST), Interleukin 6 (IL-6), tumor necrosis factor-alpha (TNF-alpha), hyaluronic acid (HA), laminin (LN), type III precollagen (PCIII), hydroxyproline (Hyp), Myeloperoxidase (MPO), malondialdehyde (MDA), superoxide dismutase (SOD), glutathione peroxidase (GSH-Px) were assayed according to the manufacturer's instructions, the alpha-SMA and TGF-beta1 were detected by western blotting, and the histopathological changes was observed by H&E staining. Isoorientin could improve the liver function by decreasing the activity of ALT, AST, IL-6, TNF-alpha, MDA, MPO, HA, LN, PCIII and Hyp (P < 0.05), increasing the activity of SOD and GSH-Px (P < 0.05), and reducing the expression of alpha-SMA and TGF-beta1 (P < 0.05). In addition, the high and middle-dose isoorientin groups showed more remarkable effect Isoorientin from G. elegans can protect hepatic fibrosis induced by alcohol.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutamic-Oxalacetic Transaminase from porcine heart, Type I, ammonium sulfate suspension, 200-500 units/mg protein
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Reagent Alcohol, denatured, suitable for HPLC
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Isoorientin, primary reference standard
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Luteolin, analytical standard
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®