MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • mRNA Expression of lipogenic enzymes in mammary tissue and fatty acid profile in milk of dairy cows fed flax hulls and infused with flax oil in the abomasum.
mRNA Expression of lipogenic enzymes in mammary tissue and fatty acid profile in milk of dairy cows fed flax hulls and infused with flax oil in the abomasum.
The British journal of nutrition (2013-11-16)
Marie-France Palin, Cristiano Côrtes, Chaouki Benchaar, Pierre Lacasse, Hélène V Petit
ABSTRACT
In the present study, the effect of flax hulls with or without flax oil bypassing the rumen on the expression of lipogenic genes in the mammary tissue of dairy cows was investigated. A total of eight dairy cows were used in a replicated 4 × 4 Latin square design. There were four periods of 21 d each and four treatments: control diet with no flax hulls (CONT); diet with 9·88 % flax hulls in the DM (HULL); control diet with 500 g flax oil/d infused in the abomasum (COFO); diet with 9·88 % flax hulls in the DM and 500 g flax oil/d infused in the abomasum (HUFO). A higher mRNA abundance of sterol regulatory element binding transcription factor, fatty acid (FA) synthase, lipoprotein lipase (LPL), PPARγ1, stearoyl-CoA desaturase (SCD) and acetyl-coenzyme A carboxylase-α was observed in cows fed HULL than in those fed CONT, and HUFO had the opposite effect. Compared with CONT, COFO and HUFO lowered the mRNA abundance of SCD, which may explain the lower proportions of MUFA in milk fat with flax oil infusion. The mRNA abundance of LPL in mammary tissue and proportions of long-chain FA in milk fat were higher in cows fed COFO than in those fed CONT. The highest proportions of trans FA were observed when cows were fed HULL. The present study demonstrates that flax hulls with or without flax oil infusion in the abomasum can affect the expression of lipogenic genes in the mammary tissue of dairy cows, which may contribute to the improvement of milk FA profile.
Sigma-Aldrich
Lipoprotein Lipase from Burkholderia sp., lyophilized powder, ≥50,000 units/mg solid
Sigma-Aldrich
Lipoprotein Lipase from Pseudomonas sp., lyophilized, powder, ≥1200 U/mg
Sigma-Aldrich
Lipoprotein Lipase from bovine milk, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)