MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Bioactivity of Piper hispidinervum (Piperales: Piperaceae) and Syzygium aromaticum (Myrtales: Myrtaceae) oils, with or without formulated Bta on the biology and immunology of Spodoptera frugiperda (Lepidoptera: Noctuidae).
Bioactivity of Piper hispidinervum (Piperales: Piperaceae) and Syzygium aromaticum (Myrtales: Myrtaceae) oils, with or without formulated Bta on the biology and immunology of Spodoptera frugiperda (Lepidoptera: Noctuidae).
Journal of economic entomology (2014-03-29)
G S Cruz, V Wanderley-Teixeira, J V Oliveira, A A Correia, M O Breda, T J S Alves, F M Cunha, A A C Teixeira, K A Dutra, D M A F Navarro
ABSTRACT
The combination of essential oils and Bacillus thuringiensis Berliner may represent an interesting control strategy. Thus, the study tested the following hypothesis: the combination of long pepper oil (Piper hispidinervum L.) and clove (Syzygium aromaticum L.) oils in two concentrations with Xentari WG (Bta) yields a more effective control of Spodoptera frugiperda (JE Smith) (Lepidoptera: Noctuidae) affecting biological and reproductive parameters and leading to changes in the levels of phenoloxidase and nitric oxide in the hemolymph of the pest. The results demonstrate that only long pepper oil, at the highest concentration with Xentari WG (Bta), promotes reduced larval survival. However, both oils with or without the insecticide interfere in the biology and humoral immunity of S.frugiperda. All treatments caused a decrease in the amount of eggs, except for the clove oil at both concentrations without Bta. Therefore, the use of these oils is a promising alternative for the integrated management of S. frugiperda; however, its association with Bta demonstrated no significant increase in their efficiency.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid