MilliporeSigma
  • Perinatal exposure to Di-(2-ethylhexyl)-Phthalate leads to cognitive dysfunction and phospho-tau level increase in aged rats.
Perinatal exposure to Di-(2-ethylhexyl)-Phthalate leads to cognitive dysfunction and phospho-tau level increase in aged rats.
Environmental toxicology (2012-05-23)
Wei Sun, Jin-Bao Ban, Ni Zhang, Yu-Kun Zu, Wen-Xiao Sun
ABSTRACT
Di-(2-ethylhexyl)-Phthalate (DEHP) can affect glucose and insulin homeostasis in periphery and lead to insulin resistance, especially exposure of DEHP during critical developmental period. Given the potential relationship between insulin resistance and pathogenesis of Alzheimer's disease (AD) in elderly life, we investigated the relationship between perinatal DEHP exposure and AD pathogenesis. Our results suggested that perinatal exposure to DEHP can affect the expression of insulin and insulin-Akt- GSK-3β signal pathway in hippocampus. Furthermore, impaired cognitive ability and increased level of phospho-Tau was observed in DEHP-exposed rat offspring (1.25 ± 0.11 vs. 0.47 ± 0.07, P < 0.05). The present study demonstrates that perinatal exposure to DEHP may be a potential risk factor for AD pathogenesis associated with insulin resistance and insulin metabolism disorder in the hippocampus.
Product Description
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland