MilliporeSigma
  • Gallic acid selectively induces the necrosis of activated hepatic stellate cells via a calcium-dependent calpain I activation pathway.
Gallic acid selectively induces the necrosis of activated hepatic stellate cells via a calcium-dependent calpain I activation pathway.
Life sciences (2014-03-19)
Shu-Chung Hsieh, Chi-Hao Wu, Chun-Chi Wu, Jung-Hsing Yen, Mei-Chun Liu, Chi-Mei Hsueh, Shih-Lan Hsu
ABSTRACT
The activation of hepatic stellate cells (HSCs) in response to liver injury is critical to the development of liver fibrosis, thus, the blockage of the activation of HSCs is considered as a rational approach for anti-fibrotic treatment. In this report, we investigated the effects and the underlying mechanisms of gallic acid (GA) in interfering with the activation of HSCs. The primary cultured rat HSCs were treated with various doses of GA for different time intervals. The morphology, viability, caspase activity, calcium ion flux, calpain I activity, reactive oxygen species generation and lysosomal functions were then investigated. GA selectively killed HSCs in both dose- and time-dependent manners, while remained no harm to hepatocytes. Besides, caspases were not involved in GA-induced cell death of HSCs. Further results showed that GA toxicity was associated with a rapid burst of reactive oxygen species (ROS) and a subsequent increase of intracellular Ca(2+) and calpain activity. Addition of calpain I but not calpain II inhibitor rescued HSCs from GA-induced death. In parallel, pretreatment with antioxidants or an intracellular Ca(2+) chelator eradicated GA responses, implying that GA-mediated cytotoxicity was dependent on its pro-oxidative properties and its effect on Ca(2+) flux. Furthermore, application of ROS scavengers also reversed Ca(2+) release and the disruption of lysosomal membranes in GA-treated HSCs. These results provide evidence for the first time that GA causes selective HSC death through a Ca(2+)/calpain I-mediated necrosis cascade, suggesting that GA may represent a potential therapeutic agent to combat liver fibrosis.
Product Description
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)