MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Carcinogenicity and other health effects of acrylonitrile with reference to occupational exposure limit.
Carcinogenicity and other health effects of acrylonitrile with reference to occupational exposure limit.
Industrial health (2000-05-17)
H Sakurai
ABSTRACT
The occupational exposure limit for acrylonitrile (AN) has been set by many organizations on the basis of its carcinogenicity. However, recent epidemiological studies do not afford evidence supporting the hypothesis that AN is carcinogenic to humans. Review of the 18 published cohort studies revealed that, although there is not adequate evidence in humans for carcinogenicity of AN, the possibility of a causal association between high exposure to AN and lung cancer in humans cannot be excluded. It was pointed out that carcinogenic potential of AN may be weak, if any, to humans, and the current occupational exposure limit (OEL) for AN of 2 ppm was evaluated as appropriate in view of AN exposure levels reported by epidemiological studies. Based also on review of the literature on health effects other than carcinogenicity, it was concluded that the current OEL for AN is a reasonable value and there is no need for a revision at present.
Product Description
Supelco
Acrylonitrile solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Acrylonitrile, analytical standard