MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Nicotinic acid: a new look at an old drug.
Current atherosclerosis reports (2009-02-21)
John A Farmer
ABSTRACT
Dyslipidemia is central to the process of atherosclerosis. Modification of the lipid profile by diet, exercise, or pharmacologic therapy has been demonstrated to reduce the risk from atherosclerosis in clinical studies in primary and secondary prevention. Nicotinic acid has been in clinical use for over 50 years. The administration of nicotinic acid has been demonstrated to reduce apolipoprotein B-containing lipoproteins (very low-density lipoprotein, intermediate-density lipoprotein, low-density lipoprotein and lipoprotein (a)). Nicotinic acid also exerts significant effects on high-density lipoprotein. In addition to improving dyslipidemia, nicotinic acid has been demonstrated to induce a number of nonlipid or pleiotropic effects. The recent discovery of the nicotinic acid receptor has improved knowledge relative to the mechanism of action and the adverse effect profile of nicotinic acid. Clinical trials utilizing clinical or angiographic end points demonstrated efficacy for the use of nicotinic acid in monotherapy or in combination with bile acid resins or statins.
Sigma-Aldrich
Nicotinic acid, SAJ special grade, ≥99.5%
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacin, United States Pharmacopeia (USP) Reference Standard
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard