MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Hydroxyurea induced dermatomyositis-like eruption.
The Australasian journal of dermatology (2012-08-14)
Tania M Zappala, Karl Rodins, James Muir
ABSTRACT
Medication-induced dermatomyositis (DM) is rare, but a recent review highlighted hydroxyurea (HU) as the most common inciting agent. To aid diagnosis, HU-induced DM-like eruption (HU DM-LE) forms a distinct dermopathy where the typical cutaneous features of DM are without systemic involvement and co-exist with other HU-induced cutaneous findings such as severe xerosis, atrophy, stomatitis, cutaneous and mucosal ulceration and melanonychia. On cessation of HU the DM-LE clears avoiding unnecessary immunosuppression and demonstrating the importance of consideration of medication aetiology in DM presentations. We present a case report and review of the literature.
Product Description
Hydroxycarbamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydroxyurea, 98%, powder