MilliporeSigma
  • Literature review on production process to obtain extra virgin olive oil enriched in bioactive compounds. Potential use of byproducts as alternative sources of polyphenols.
Literature review on production process to obtain extra virgin olive oil enriched in bioactive compounds. Potential use of byproducts as alternative sources of polyphenols.
Journal of agricultural and food chemistry (2013-05-10)
Edwin Frankel, Abdelhakim Bakhouche, Jesús Lozano-Sánchez, Antonio Segura-Carretero, Alberto Fernández-Gutiérrez
ABSTRACT
This review describes the olive oil production process to obtain extra virgin olive oil (EVOO) enriched in polyphenol and byproducts generated as sources of antioxidants. EVOO is obtained exclusively by mechanical and physical processes including collecting, washing, and crushing of olives, malaxation of olive paste, centrifugation, storage, and filtration. The effect of each step is discussed to minimize losses of polyphenols from large quantities of wastes. Phenolic compounds including phenolic acids, alcohols, secoiridoids, lignans, and flavonoids are characterized in olive oil mill wastewater, olive pomace, storage byproducts, and filter cake. Different industrial pilot plant processes are developed to recover phenolic compounds from olive oil byproducts with antioxidant and bioactive properties. The technological information compiled in this review will help olive oil producers to improve EVOO quality and establish new processes to obtain valuable extracts enriched in polyphenols from byproducts with food ingredient applications.
Product Description
Supelco
Olive oil, analytical standard
Sigma-Aldrich
Olive oil, highly refined, low acidity
Sigma-Aldrich
Olive oil, tested according to Ph. Eur.