MilliporeSigma
  • Synthesis and characterization of some new 4-hydroxy-coumarin derivatives.
Synthesis and characterization of some new 4-hydroxy-coumarin derivatives.
Molecules (Basel, Switzerland) (2014-08-12)
Yasameen K Al-Majedy, Abdul Amir H Kadhum, Ahmed A Al-Amiery, Abu Bakar Mohamad
ABSTRACT
Some novel coumarins were synthesized starting from 4-hydroxycoumarin and methyl bromoacetate. The structures of the newly obtained compounds were confirmed by elemental analysis, mass, IR and NMR spectra.
Sigma-Aldrich
Methyl bromoacetate, 97%
Pricing and availability is not currently available.