MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Revealing organic carbon-nitrate linear relationship from UV spectra of freshwaters in agricultural environment.
Revealing organic carbon-nitrate linear relationship from UV spectra of freshwaters in agricultural environment.
Chemosphere (2014-05-31)
O Thomas, A V Jung, J Causse, M V Louyer, S Piel, E Baurès, M F Thomas
ABSTRACT
A strong non linear relationship between nitrate and organic matter (assessed by dissolved organic carbon, DOC) has been recently demonstrated by Taylor and Townsend (2010), namely for freshwaters. In this context, our study explores this relation from the behavior of sets of normalized UV spectra (same area under each spectrum) of different water samples showing a hidden isosbestic point (HIP) around 225 nm. This HIP is linked to the existence of a simple relation between nitrate and DOC, the proportions of which vary according to the sampling location and environmental factors. In a second step, a simple linear model is proposed for nitrate-DOC relationship (α⋅NO3+β⋅DOC=1) and a validation is proposed for more than 150 samples of different Brittany rivers and lakes. For samples of the largest watershed, a complementary exploitation from data acquired during the different campaigns confirmed the seasonal evolution between spring (high nitrate/low DOC) and autumn (high DOC/low nitrate). Further investigation on other freshwater samples is needed in order to improve the limits of this linear model.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%