MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Characterization of the interaction of diacylglycerol acyltransferase-2 with the endoplasmic reticulum and lipid droplets.
Characterization of the interaction of diacylglycerol acyltransferase-2 with the endoplasmic reticulum and lipid droplets.
Biochimica et biophysica acta (2014-06-24)
Pamela J McFie, Youzhi Jin, Shanna L Banman, Erwan Beauchamp, Luc G Berthiaume, Scot J Stone
ABSTRACT
Acyl CoA:diacylglycerol acyltransferase-2 (DGAT2) is an integral membrane protein that catalyzes the synthesis of triacylglycerol (TG). DGAT2 is present in the endoplasmic reticulum (ER) and also localizes to lipid droplets when cells are stimulated with oleate. Previous studies have shown that DGAT2 can interact with membranes and lipid droplets independently of its two transmembrane domains, suggesting the presence of an additional membrane binding domain. In order to identify additional membrane binding regions, we confirmed that DGAT2 has only two transmembrane domains and demonstrated that the loop connecting them is present in the ER lumen. Increasing the length of this short loop from 5 to 27 amino acids impaired the ability of DGAT2 to localize to lipid droplets. Using a mutagenesis approach, we were able to identify a stretch of amino acids that appears to have a role in binding DGAT2 to the ER membrane. Our results confirm that murine DGAT2 has only two transmembrane domains but also can interact with membranes via a previously unidentified helical domain containing its active site.
Product Description
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%