MilliporeSigma
  • Tubular calcium reabsorption and other aspects of calcium homeostasis in primary and secondary hyperparathyroidism.
Tubular calcium reabsorption and other aspects of calcium homeostasis in primary and secondary hyperparathyroidism.
Clinical nephrology (2014-07-06)
Kenneth R Phelps, Kim S Stote, Darius Mason
ABSTRACT
Primary hyperparathyroidism (PHPT) causes hypercalcemia by increasing tubular calcium reabsorption. Because chronic kidney disease (CKD) is associated with normocalcemia, we inferred that calcium reabsorption is also normal, and hypothesized that normal reabsorption requires excessive parathyroid hormone (PTH) in CKD. The following were obtained in controls and patients with CKD or PHPT: estimated GFR (eGFR); concentrations of PTH 1-84, 1,25-dihydroxyvitamin D, and ultrafilterable and ionized calcium ([PTH], [1,25(OH)2D], [Ca]uf, [Ca]i); and ratios of calcium excreted or reabsorbed per volume of filtrate (ECa/Ccr, TRCa/Ccr). Pertinent linear regressions were examined. In CKD, [PTH] was increased, but ECa/Ccr, TRCa/Ccr, [Ca]uf, and [Ca]i equaled control values. [PTH] was inversely related to eGFR but unrelated to [1,25(OH)2D]. TRCa/Ccr was constant at all [PTH]. In PHPT, [PTH] was no higher than in CKD, but TRCa/Ccr, [Ca]uf, and [Ca]i were increased. [1,25(OH)2D] correlated with [PTH]. In controls, TRCa/Ccr varied directly with [1,25(OH)2D] and inversely with [PTH]. In controls, calcium reabsorption rose with [1,25(OH)2D], and [PTH] fell in response. In PHPT, [PTH] determined [1,25(OH)2D]; together, the hormones increased calcium reabsorption and caused hypercalcemia. In CKD, normal calcium reabsorption required high [PTH].
Sigma-Aldrich
Calcium, SAJ first grade, ≥99.5%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%