MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The Trichoplusia ni single nucleopolyhedrovirus tn79 gene encodes a functional sulfhydryl oxidase enzyme that is able to support the replication of Autographa californica multiple nucleopolyhedrovirus lacking the sulfhydryl oxidase ac92 gene.
The Trichoplusia ni single nucleopolyhedrovirus tn79 gene encodes a functional sulfhydryl oxidase enzyme that is able to support the replication of Autographa californica multiple nucleopolyhedrovirus lacking the sulfhydryl oxidase ac92 gene.
Virology (2014-07-11)
Stian A Clem, Wenbi Wu, A Lorena Passarelli
ABSTRACT
The Autographa californica multiple nucleopolyhedrovirus ac92 is a conserved baculovirus gene with homology to flavin adenine dinucleotide-linked sulfhydryl oxidases. Its product, Ac92, is a functional sulfhydryl oxidase. Deletion of ac92 results in almost negligible levels of budded virus (BV) production, defects in occlusion-derived virus (ODV) co-envelopment and their inefficient incorporation into occlusion bodies. To determine the role of sulfhydryl oxidation in the production of BV, envelopment of nucleocapsids, and nucleocapsid incorporation into occlusion bodies, the Trichoplusia ni single nucleopolyhedrovirus ortholog, tn79, was substituted for ac92. Tn79 was found to be an active sulfhydryl oxidase that substituted for Ac92, resulting in the production of infectious BV, albeit about 10-fold less than an ac92-containing virus. Tn79 rescued defects in ODV morphogenesis caused by a lack of ac92. Active Tn79 sulfhydryl oxidase activity is required for efficient BV production, ODV envelopment, and their subsequent incorporation into occlusion bodies in the absence of ac92.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Hypromellose, meets USP testing specifications
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000