MilliporeSigma
  • Role of Template Activating Factor-I as a chaperone in linker histone dynamics.
Role of Template Activating Factor-I as a chaperone in linker histone dynamics.
Journal of cell science (2011-09-24)
Kohsuke Kato, Mitsuru Okuwaki, Kyosuke Nagata
ABSTRACT
Linker histone H1 is a fundamental chromosomal protein involved in the maintenance of higher-ordered chromatin organization. The exchange dynamics of histone H1 correlates well with chromatin plasticity. A variety of core histone chaperones involved in core histone dynamics has been identified, but the identity of the linker histone chaperone in the somatic cell nucleus has been a long-standing unanswered question. Here we show that Template Activating Factor-I (TAF-I, also known as protein SET) is involved in histone H1 dynamics as a linker histone chaperone. Among previously identified core histone chaperones and linker histone chaperone candidates, only TAF-I was found to be associated specifically with histone H1 in mammalian somatic cell nuclei. TAF-I showed linker histone chaperone activity in vitro. Fluorescence recovery after photobleaching analyses revealed that TAF-I is involved in the regulation of histone H1 dynamics in the nucleus. Therefore, we propose that TAF-I is a key molecule that regulates linker histone-mediated chromatin assembly and disassembly.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.