MilliporeSigma
A systematic review of peri-operative melatonin.
Anaesthesia (2014-05-20)
L P H Andersen, M U Werner, J Rosenberg, I Gögenur
ABSTRACT
We systematically reviewed randomised controlled trials of peri-operative melatonin. We included 24 studies of 1794 participants that reported eight peri-operative outcomes: anxiety; analgesia; sleep quality; oxidative stress; emergence behaviour; anaesthetic requirements; steal induction; and safety. Compared with placebo, melatonin reduced the standardised mean difference (95% CI) pre-operative anxiety score by 0.88 (0.44-1.33) and postoperative pain score by 1.06 (0.23-1.88). The magnitude of effect was unreliable due to substantial statistical heterogeneity, with I(2) 87% and 94%, respectively. Qualitative reviews suggested the melatonin improved sleep quality and emergence behaviour, and might be capable of reducing oxidative stress and anaesthetic requirements.
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
USP
Melatonin, United States Pharmacopeia (USP) Reference Standard