MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Non-steroidal anti-inflammatory drugs and cancer risk in women: results from the Women's Health Initiative.
Non-steroidal anti-inflammatory drugs and cancer risk in women: results from the Women's Health Initiative.
International journal of cancer (2014-03-07)
Theodore M Brasky, Jingmin Liu, Emily White, Ulrike Peters, John D Potter, Roland B Walter, Christina S Baik, Dorothy S Lane, JoAnn E Manson, Mara Z Vitolins, Matthew A Allison, Jean Y Tang, Jean Wactawski-Wende
ABSTRACT
The use of non-steroidal anti-inflammatory drugs (NSAIDs) has been associated with reduced risks of cancers at several sites in some studies; however, we recently reported no association between their use and total cancer risk in women in a prospective study. Here we examine the association between NSAIDs and total and site-specific cancer incidence in the large, prospective Women's Health Initiative (WHI). Women (129,013) were recruited to participate in the WHI at 40 US clinical centers from 1993 to 1998 and followed prospectively. After 9.7 years of follow-up, 12,998 incident, first primary, invasive cancers were diagnosed. NSAID use was systematically collected at study visits. We used Cox proportional hazards regression models to estimate multivariable-adjusted hazard ratios (HR) and 95% confidence intervals (CI) for associations between NSAIDs use and total and site-specific cancer risk. Relative to non-use, consistent use (i.e., use at baseline and year 3 of follow-up) of any NSAID was not associated with total cancer risk (HR 1.00, 95% CI: 0.94-1.06). Results for individual NSAIDs were similar to the aggregate measure. In site-specific analyses, NSAIDs were associated with reduced risks of colorectal cancer, ovarian cancer, and melanoma. Our study confirms a chemopreventive benefit for colorectal cancer in women and gives preliminary evidence for a reduction of the risk of some rarer cancers. NSAIDs' benefit on cancer risk was therefore limited to specific sites and not evident when total cancer risk was examined. This information may be of importance when NSAIDs are considered as chemopreventive agents.
Product Description
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Naproxen, European Pharmacopoeia (EP) Reference Standard
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Naproxen, VETRANAL®, analytical standard
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard